missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorobenzaldehyde, 97%

Product Code. 11348476
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348476 5 g 5g
11338476 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348476 Supplier Thermo Scientific Alfa Aesar Supplier No. L17515.06
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20028-53-9 | C7H6ClNO | 155.581 g/mol

2-Amino-5-Chlorobenzaldehyde is used as a raw material in organic synthesis. It also serves as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-Chlorobenzaldehyde is used as a raw material in organic synthesis. It also serves as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20028-53-9
Molecular Formula C7H6ClNO
Molecular Weight (g/mol) 155.581
MDL Number MFCD02093936
InChI Key KUIFMGITZFDQMP-UHFFFAOYSA-N
Synonym pubchem7364, acmc-209vm9, 3-chloro-6-aminobenzaldehyde, 2-amino-5-chloro-benzaldehyde, 2-amino-5-chlorobenz-aldehyde, 5-chloroanthranilaldehyde
PubChem CID 2773898
IUPAC Name 2-amino-5-chlorobenzaldehyde
SMILES C1=CC(=C(C=C1Cl)C=O)N

Specifications

Melting Point 68°C to 74°C
Quantity 5 g
Beilstein 2716096
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 155.58
Percent Purity 97%
Chemical Name or Material 2-Amino-5-chlorobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.