missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromopyrimidine, 97%

Product Code. 11362349
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362349 5 g 5g
11352349 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362349 Supplier Thermo Scientific Alfa Aesar Supplier No. H27291.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7752-82-1 | C4H4BrN3 | 174.001 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7752-82-1
Molecular Formula C4H4BrN3
Molecular Weight (g/mol) 174.001
MDL Number MFCD00012341
InChI Key UHRHPPKWXSNZLR-UHFFFAOYSA-N
Synonym 2-amino-5-bromopyrimidine, 5-bromopyrimidin-2-ylamine, 2-pyrimidinamine, 5-bromo, 5-bromo-2-aminopyrimidine, 5-bromo-2-pyrimidinamine, pyrimidine, 2-amino-5-bromo, 5-bromopyrimidine-2-amine, 2-amino-5-bromo-pyrimidine, 5-bromopyrimidine-2-ylamine, 5-bromo-pyrimidin-2-ylamine
PubChem CID 231310
IUPAC Name 5-bromopyrimidin-2-amine
SMILES C1=C(C=NC(=N1)N)Br

Specifications

Melting Point 232°C to 234°C
Quantity 5 g
Formula Weight 174
Percent Purity 97%
Chemical Name or Material 2-Amino-5-bromopyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.