missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromopyridine, 98%

Product Code. 11482437
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11482437 100 g 100g
11462437 5 g 5g
11472437 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482437 Supplier Thermo Scientific Alfa Aesar Supplier No. A14280.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1072-97-5 | C5H5BrN2 | 173.013 g/mol

2-Amino-5-bromopyridine is used for the synthesis of polycyclic azaarenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-bromopyridine is used for the synthesis of polycyclic azaarenes.

Solubility
Soluble in methanol, chloroform, ethyl acetate. Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Light sensitive. Store under argon.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-97-5
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.013
MDL Number MFCD00006323
InChI Key WGOLHUGPTDEKCF-UHFFFAOYSA-N
Synonym 2-amino-5-bromopyridine, 2-pyridinamine, 5-bromo, 5-bromo-2-pyridylamine, 5-bromo-2-aminopyridine, 5-bromo-2-pyridinamine, 5-bromopyridin-2-ylamine, unii-d5qe8xw52u, 5-bromo-pyridin-2-ylamine, 2-amino-5-bromo pyridine, d5qe8xw52u
PubChem CID 70622
IUPAC Name 5-bromopyridin-2-amine
SMILES C1=CC(=NC=C1Br)N

Specifications

Melting Point 135°C to 140°C
Quantity 100 g
Beilstein 108737
Solubility Information Soluble in methanol,chloroform,ethyl acetate. Slightly soluble in water.
Formula Weight 173.01
Percent Purity 98%
Chemical Name or Material 2-Amino-5-bromopyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.