missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromopyridine, 97%

Product Code. 10163440 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10163440 25 g 25g
10347900 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10163440 Supplier Thermo Scientific Acros Supplier No. 153740250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1072-97-5 | C5H5BrN2 | 173.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-97-5
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.01
MDL Number MFCD00006323
InChI Key WGOLHUGPTDEKCF-UHFFFAOYSA-N
Synonym 2-amino-5-bromopyridine, 2-pyridinamine, 5-bromo, 5-bromo-2-pyridylamine, 5-bromo-2-aminopyridine, 5-bromo-2-pyridinamine, 5-bromopyridin-2-ylamine, unii-d5qe8xw52u, 5-bromo-pyridin-2-ylamine, 2-amino-5-bromo pyridine, d5qe8xw52u
PubChem CID 70622
IUPAC Name 5-bromopyridin-2-amine
SMILES C1=CC(=NC=C1Br)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 135.0°C to 138.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula Br(C5H3N)NH2
Quantity 25 g
Beilstein 22, 431
Formula Weight 173.01
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-bromopyridine, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.