missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromobenzaldehyde, 97%

Product Code. 15496297
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15496297 1 g 1g
15406307 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496297 Supplier Thermo Scientific Alfa Aesar Supplier No. H33491.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 29124-57-0 | C7H6BrNO | 200.035 g/mol

Participate in synthesis and structure activity of novel camptothecin analogs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Participate in synthesis and structure activity of novel camptothecin analogs.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29124-57-0
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.035
MDL Number MFCD10696879
InChI Key VBYZWJMZASVGNB-UHFFFAOYSA-N
Synonym 2-amino-5-bromobenzenecarbaldehyde, 5-bromoanthranilaldehyde, 4-bromo-2-formylaniline, benzaldehyde,2-amino-5-bromo, benzaldehyde, 2-amino-5-bromo, pubchem19693, aminobromobenzenecarbaldehyde, 2-amino-5-bromobenzaidehyde, 2-amino-5-bromo-benzaldehyde
PubChem CID 23510475
IUPAC Name 2-amino-5-bromobenzaldehyde
SMILES C1=CC(=C(C=C1Br)C=O)N

Specifications

Melting Point 75°C to 77°C
Color Yellow
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 200
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Amino-5-bromobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.