missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-3-pyridinemethanol, ≥98%, Thermo Scientific™

Catalog Number p-7055807
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
335031-01-1
C6H7BrN2O
203.039
MFCD05664025
HIVQAWXDBPYNDU-UHFFFAOYSA-N
2-amino-5-bromo-3-hydroxymethyl pyridine, 2-amino-5-bromopyridin-3-yl methanol, 2-amino-5-bromo-3-pyridinemethanol, 2-amino-5-bromopyridine-3-methanol, 2-amino-5-bromo-3-pyridinyl methanol, 2-amino-5-bromo-pyridin-3-yl-methanol, 3-pyridinemethanol, 2-amino-5-bromo, 2-amino-5-bromo-3-pyridyl methan-1-ol, 2-amino-5-bromopyridin-3-yl-methanol, pubchem9344
10891366
(2-amino-5-bromopyridin-3-yl)methanol
C1=C(C(=NC=C1Br)N)CO
This item is not returnable. View return policy
335031-01-1
C6H7BrN2O
203.039
MFCD05664025
HIVQAWXDBPYNDU-UHFFFAOYSA-N
2-amino-5-bromo-3-hydroxymethyl pyridine, 2-amino-5-bromopyridin-3-yl methanol, 2-amino-5-bromo-3-pyridinemethanol, 2-amino-5-bromopyridine-3-methanol, 2-amino-5-bromo-3-pyridinyl methanol, 2-amino-5-bromo-pyridin-3-yl-methanol, 3-pyridinemethanol, 2-amino-5-bromo, 2-amino-5-bromo-3-pyridyl methan-1-ol, 2-amino-5-bromopyridin-3-yl-methanol, pubchem9344
10891366
(2-amino-5-bromopyridin-3-yl)methanol
C1=C(C(=NC=C1Br)N)CO

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White to Yellow
Quantity 1 g
Beilstein 9387621
Formula Weight 203.04
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material 2-Amino-5-bromo-3-pyridinemethanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'