missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-3-picoline, 97%, Thermo Scientific™

Product Code. 15302032 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15302032 10 g 10g
15312032 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15302032 Supplier Thermo Scientific Acros Supplier No. 360880100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 3430-21-5
Molecular Formula C6H7BrN2
Molecular Weight (g/mol) 187.04
InChI Key KBLGGRWUEVCNPY-UHFFFAOYSA-N
Synonym 2-amino-5-bromo-3-methylpyridine, 2-amino-5-bromo-3-picoline, 2-amino-3-methyl-5-bromopyridine, 5-bromo-3-methyl-pyridin-2-ylamine, 2-pyridinamine, 5-bromo-3-methyl, 5-bromo-3-methyl-2-pyridinamine, 2-amina-5-bromo-3-picoline, 5-bromo-3-methyl-2-pyridylamine, 5-bromo-3-methyl-pyridin-2-amine, 5-bromo-3-methylpyridin-2-ylamine
PubChem CID 137934
IUPAC Name 5-bromo-3-methylpyridin-2-amine
SMILES CC1=CC(=CN=C1N)Br

Specifications

Melting Point 92.0°C to 94.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 10 g
Solubility Information Solubility in water: slighlty soluble. Other solubilities: soluble in methanol
Formula Weight 187.04
Percent Purity 97%
Chemical Name or Material 2-Amino-5-bromo-3-picoline
compliance-icons
Product Identifier
  • 2-Amino-5-bromo-3-picoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.