missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-3-hydroxypyridine, 95%, Thermo Scientific Chemicals

Product Code. 15418288
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15418288 1 g 1g
15428288 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418288 Supplier Thermo Scientific Alfa Aesar Supplier No. H61722.03
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 39903-01-0 | Molecular Formula: C5H5BrN2O | Molecular Weight: 189.01 g/mol

2-Amino-5-bromo-3-hydroxypyridine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-bromo-3-hydroxypyridine is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39903-01-0
Molecular Formula C5H5BrN2O
Molecular Weight (g/mol) 189.01
MDL Number MFCD09744143
InChI Key YQADLKDQAXAIKW-UHFFFAOYSA-N
Synonym 2-amino-5-bromo-3-pyridinol, 2-amino-5-bromo-3-hydroxypyridine, 2-amino-3-hydroxy-5-bromopyridine, 3-pyridinol, 2-amino-5-bromo, 2-amino-5-bromo-pyridin-3-ol, 2-amino-5-bromopyridine-3-ol, acmc-209j8w, 2-amino-5-bromopryidin-3-ol, 2-amino-3-hydroxy-5-bromo pyridine, 5-bromo-3-hydroxy-2-amino-pyridine
PubChem CID 11694041
IUPAC Name 2-amino-5-bromopyridin-3-ol
SMILES NC1=NC=C(Br)C=C1O

Specifications

Color White
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 189.01
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Amino-5-bromo-3-hydroxypyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.