missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile

Product Code. 15497975
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15497975

Brand: Thermo Scientific Alfa Aesar H32059.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 70291-62-2 | C8H8N2S | 164.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70291-62-2
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.23
MDL Number MFCD00121495
InChI Key RILAXFOADRDGPQ-UHFFFAOYSA-N
PubChem CID 258182
IUPAC Name 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile
SMILES NC1=C(C#N)C2=C(CCC2)S1

Specifications

Melting Point 153°C to 154°C
Quantity 5 g
UN Number UN3439
Synonym 2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-carbonitrile, 2-amino-4h,5h,6h-cyclopenta b thiophene-3-carbonitrile, 2-amino-4,5-trimethylene-3-thiophenecarbonitrile, 4h-cyclopenta b thiophene-3-carbonitrile, 2-amino-5,6-dihydro, 2-amino-3-cyano-5,6-dihydro-4h-cyclopenta b thiophene, 2-amino-4,5,6-trihydrocyclopenta 1,2-b thiophene-3-carbonitrile, 4h-cyclopenta b thiophene-3-carbonitrile,2-amino-5,6-dihydro, maybridge1_003206, 4,5-trimethylene-2-aminothiophene-3-carbonitrile, 2-amino-5,6-dihydro-4h-cyclopenta b thioph ene-3-carbonitrile
Formula Weight 164.23
Chemical Name or Material 2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.