missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(trifluoromethyl)thiophenol hydrochloride, 97%

Product Code. 11372207
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372207 5 g 5g
11362207 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372207 Supplier Thermo Scientific Alfa Aesar Supplier No. L09683.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4274-38-8 | C7H7ClF3NS | 229.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4274-38-8
Molecular Formula C7H7ClF3NS
Molecular Weight (g/mol) 229.65
MDL Number MFCD00042600
InChI Key FIAGYDIJZOWVAB-UHFFFAOYSA-N
Synonym 2-amino-4-trifluoromethyl benzenethiol hydrochloride, 3-amino-4-mercaptobenzotrifluoride hydrochloride, 2-amino-4-trifluoromethylthiophenol hydrochloride, 2-amino-4-trifluoromethyl benzene-1-thiol hydrochloride, 2-amino-4-trifluoromethyl thiophenol hydrochloride, 2-amino-4-trifluoromethyl benzene-1-thiol, chloride, pubchem2827, c7h6f3ns.hcl, dsstox_cid_31681, dsstox_rid_97532
PubChem CID 2734678
IUPAC Name 2-amino-4-(trifluoromethyl)benzenethiol;hydrochloride
SMILES [H+].[Cl-].NC1=CC(=CC=C1S)C(F)(F)F

Specifications

Melting Point 186°C to 190°C
Density 2.46
Quantity 5 g
UN Number UN1759
Beilstein 4273774
Formula Weight 229.65
Percent Purity 97%
Chemical Name or Material 2-Amino-4-(trifluoromethyl)thiophenol hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.