missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(trifluoromethyl)pyridine, 99%, Thermo Scientific Chemicals

Product Code. 11378936
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378936 500 mg 500mg
11388936 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378936 Supplier Thermo Scientific Alfa Aesar Supplier No. L19548.ME
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106447-97-6
Molecular Formula C6H5F3N2
Molecular Weight (g/mol) 162.115
MDL Number MFCD00077455
InChI Key RWGBXAQMUBGGKQ-UHFFFAOYSA-N
Synonym 2-amino-4-trifluoromethyl pyridine, 4-trifluoromethyl pyridin-2-amine, 4-trifluoromethyl-pyridin-2-ylamine, 2-amino-4-trifluoromethylpyridine, 4-trifluoromethyl-2-pyridinamine, 4-trifluoromethyl-2-pyridylamine, 2-pyridinamine, 4-trifluoromethyl, 2-amino-4-trifluoromethyl-pyridine, pubchem2977
PubChem CID 2778315
IUPAC Name 4-(trifluoromethyl)pyridin-2-amine
SMILES C1=CN=C(C=C1C(F)(F)F)N

Specifications

Melting Point 71°C to 74°C
Quantity 500 mg
UN Number UN2811
Beilstein 7869677
Formula Weight 162.11
Percent Purity 99%
Chemical Name or Material 2-Amino-4-(trifluoromethyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.