missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester, 95%

Product Code. 15471106
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471106 250 mg 250mg
15461106 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15471106 Supplier Thermo Scientific Alfa Aesar Supplier No. H54372.MD
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 944401-57-4 | C12H16BF3N2O2 | 288.077 g/mol

2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 944401-57-4
Molecular Formula C12H16BF3N2O2
Molecular Weight (g/mol) 288.077
MDL Number MFCD12923420
InChI Key AHNBKJSRXQDYEO-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-4-trifluoromethyl pyridin-2-amine, 2-amino-4-trifluoromethyl pyridine-5-boronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-4-trifluoromethyl pyridin-2-amine, 6-amino-4-trifluoromethyl pyridin-3-yl boronic acid pinacol ester, 4-trifluoromethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, 4-trifluoromethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridm-2-amine, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-4-trifluoromethyl pyridin-2-amine
PubChem CID 57416499
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)pyridin-2-amine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2C(F)(F)F)N

Specifications

Quantity 250 mg
Beilstein 15519102
Solubility Information Slightly soluble in water.
Formula Weight 288.07
Percent Purity 95%
Chemical Name or Material 2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.