Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifica
| CAS Min % | 96.0 |
| CAS Max % | 100.0 |
| Melting Point | 150.0°C to 152.5°C |
| Color | Orange to White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (HPLC) |
| Packaging | Glass bottle |
| Quantity | 5 g |
| Synonym | 2-benzyloxy-3-methoxybenzaldehyde, 2-benzyloxy-3-methoxy-benzaldehyde, 3-methoxy-2-phenylmethoxy benzaldehyde, acmc-1cljo, benzyloxy-3-methoxy benzaldehyde, 2-benzyloxy-3-methoxyl-benzaldehyde, 2-benzyloxy-3-methoxybenzenecarbaldehyde |
| SMILES | NC1=NC(=CS1)C1=CC=CC=C1 |
| Vedi altri risultati |
Sicurezza e movimentazione
- 2-Amino-4-phenylthiazole
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.