missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-phenylthiazole, 97%, Thermo Scientific Chemicals

Catalog Number p-4228339
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2010-06-2
C9H8N2S
176.24
MFCD00039680
PYSJLPAOBIGQPK-UHFFFAOYSA-N
This item is not returnable. View return policy
2010-06-2
C9H8N2S
176.24
MFCD00039680
PYSJLPAOBIGQPK-UHFFFAOYSA-N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 150.0°C to 152.5°C
Color Orange to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Synonym 4-phenylthiazol-2-amine, 2-amino-4-phenylthiazole, phenthiazamine, 2-thiazolamine, 4-phenyl, 4-phenyl-2-thiazolamine, thiazole, 2-amino-4-phenyl, 4-phenyl-thiazol-2-ylamine, 4-phenyl-2-thiazolylamine, 2-amino-4-phenyl-1,3-thiazole, 4-phenyl-2-aminothiazole
SMILES C1=CC=C(C=C1)C2=CSC(=N2)N
IUPAC Name 4-phenyl-1,3-thiazol-2-amine
PubChem CID 40302
Formula Weight 176.24
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 2-Amino-4-phenylthiazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Amino-4-phenylthiazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'