missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylpyridine, 98%

Product Code. 11472237
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472237 500 g 500g
11349689 25 g 25g
11462237 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472237 Supplier Thermo Scientific Alfa Aesar Supplier No. A14177.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 695-34-1 | C6H8N2 | 108.144 g/mol

2-Amino-4-methylpyridine has been used in the synthesis of 2-amino-4-methyl­pyridinium 2-hy­droxy­benzoate. It is potent inhibitor of NOS2 (iNOS) in vitro. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-methylpyridine has been used in the synthesis of 2-amino-4-methyl­pyridinium 2-hy­droxy­benzoate. It is potent inhibitor of NOS2 (iNOS) in vitro. It is used as pharmaceutical intermediate.

Solubility
Soluble in water (41 mg/ml at 20°C), lower alcohols (slightly), chloroform, ethyl acetate, and DMF.

Notes
Hygroscopic. Incompatible with oxidizing agents and moisture. Store under dry inert gas. Protect from humidity and water. Store at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 695-34-1
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006321
InChI Key ORLGLBZRQYOWNA-UHFFFAOYSA-N
Synonym 2-amino-4-methylpyridine, 2-amino-4-picoline, 2-pyridinamine, 4-methyl, ascensil, 4-methyl-2-pyridinamine, 4-methyl-2-pyridylamine, 4-methyl-2-aminopyridine, 4-picoline, 2-amino, w 45 raschig, aminton
PubChem CID 1533
IUPAC Name 4-methylpyridin-2-amine
SMILES CC1=CC(=NC=C1)N

Specifications

Melting Point 97°C to 101°C
Boiling Point 231°C
Flash Point 118°C (244°F)
Odor Unpleasant
Quantity 500 g
UN Number UN2811
Beilstein 107066
Sensitivity Hygroscopic
Merck Index 14,466
Solubility Information Soluble in water (41mg/ml at 20°C),lower alcohols (slightly),chloroform,ethyl acetate,and DMF.
Formula Weight 108.14
Percent Purity 98%
Chemical Name or Material 2-Amino-4-methylpyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.