missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylphenol, 98%

Product Code. 11454117
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454117 250 g 250g
11444117 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454117 Supplier Thermo Scientific Alfa Aesar Supplier No. A19546.30
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 95-84-1 | C7H9NO | 123.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-84-1
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00007699
InChI Key ZMXYNJXDULEQCK-UHFFFAOYSA-N
Synonym 2-amino-p-cresol, 2-hydroxy-5-methylaniline, phenol, 2-amino-4-methyl, 3-amino-4-hydroxytoluene, 6-hydroxy-m-toluidine, 4-methyl-2-aminophenol, 5-methyl-2-hydroxyaniline, 2-amino-4-cresol, p-cresol, 2-amino, 2-amino-4-methyl-phenol
PubChem CID 7264
IUPAC Name 2-amino-4-methylphenol
SMILES CC1=CC=C(O)C(N)=C1

Specifications

Melting Point 134°C to 138°C
Quantity 250 g
Beilstein 606494
Formula Weight 123.16
Percent Purity 98%
Chemical Name or Material 2-Amino-4-methylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.