missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylbenzothiazole, 98%

Product Code. 11419772
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419772 5 g 5g
11429772 25 g 25g
11439772 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11419772 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' A10822.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1477-42-5 | C8H8N2S | 164.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1477-42-5
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.23
MDL Number MFCD00005793
InChI Key GRIATXVEXOFBGO-UHFFFAOYSA-N
Synonym 2-amino-4-methylbenzothiazole, 4-methylbenzo d thiazol-2-amine, 2-benzothiazolamine, 4-methyl, 4-methyl-2-aminobenzothiazole, 4-methyl-2-benzothiazolamine, benzothiazole, 2-amino-4-methyl, 4-methylbenzothiazol-2-ylamine, 2-amino-4-methyl benzothiazole, 4-methyl-2-aminobenzothiozole, unii-45v5f1b9z8
PubChem CID 15132
IUPAC Name 4-methyl-1,3-benzothiazol-2-amine
SMILES CC1=C2N=C(N)SC2=CC=C1

Specifications

Melting Point 137°C to 139°C
Quantity 5 g
Beilstein 129346
Sensitivity Moisture sensitive
Formula Weight 164.22
Percent Purity 98%
Chemical Name or Material 2-Amino-4-methylbenzothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.