missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylbenzonitrile, 97%, Thermo Scientific™

Product Code. p-2697214
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10672273

Brand: Thermo Scientific Maybridge TL00552DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 26830-96-6
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD00173706
InChI Key LGNVAEIITHYWCG-UHFFFAOYSA-N
Synonym 2-cyano-5-methylaniline, 3-amino-4-cyanotoluene, 2-amino-4-methyl-benzonitrile, 4-methyl-2-aminobenzonitrile, benzonitrile, 2-amino-4-methyl, 2-amino-4-methylbenzenecarbonitrile, 2-amino-4-methylbenzonitril, 6-cyano-m-toluidine, pubchem4661, 4-methylanthranilonitrile
PubChem CID 2801276
IUPAC Name 2-amino-4-methylbenzonitrile
SMILES CC1=CC=C(C#N)C(N)=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 132.17
Percent Purity 97%
Chemical Name or Material 2-Amino-4-methylbenzonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.