missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylbenzoic acid, 95%, Thermo Scientific™

Product Code. 10170802
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
10 g
missing translation for 'unitSize'
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10170802 1 g 1g
10118474 5 g 5g
10445912 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10170802 missing translation for 'mfr' Thermo Scientific Maybridge missing translation for 'supplierNo' TL00555DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 2305-36-4
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00047853
InChI Key RPGKFFKUTVJVPY-UHFFFAOYSA-N
Synonym 4-methylanthranilic acid, benzoic acid, 2-amino-4-methyl, 2-amino-4-methyl-benzoic acid, 2-amino-p-toluic acid, 3-amino-4-carboxytoluene, 2-carboxy-5-methylaniline, p-toluic acid, 2-amino, rarechem al bo 0820, benzoic acid, 2-amino-4-methyl-9ci, 2-azanyl-4-methyl-benzoic acid
PubChem CID 75316
IUPAC Name 2-amino-4-methylbenzoic acid
SMILES CC1=CC(=C(C=C1)C(=O)O)N

Specifications

CAS Min % 97.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 151.16
Percent Purity 97%
Chemical Name or Material 2-Amino-4-methylbenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.