missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4'-methoxyacetophenone hydrochloride, 97%

Product Code. 10441674 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10441674 1 g Glass bottle 1g
10071034 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10441674 Supplier Thermo Scientific Acros Supplier No. 425710010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3883-94-1 | C9H11NO2·ClH | 201.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3883-94-1
Molecular Formula C9H11NO2·ClH
Molecular Weight (g/mol) 201.65
MDL Number MFCD00193078
InChI Key FZVYWBMMOSHMRS-UHFFFAOYSA-N
Synonym 2-amino-1-4-methoxyphenyl ethanone hydrochloride, 2-amino-4'-methoxyacetophenone hydrochloride, 2-amino-1-4-methoxyphenyl ethan-1-one hydrochloride, 4-methoxyphenacylamine hydrochloride, ethanone, 2-amino-1-4-methoxyphenyl-, hydrochloride, 2-amino-1-4-methoxyphenyl ethan-1-one, chloride, pubchem22350, acmc-1ad40, 4-aminoacetyl anisole hydrochloride, 2-amino-4-methoxyacetophenone hydrochloride
PubChem CID 12487188
IUPAC Name 2-amino-1-(4-methoxyphenyl)ethanone;hydrochloride
SMILES COC1=CC=C(C=C1)C(=O)CN.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 190°C to 193°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3OC6H4COCH2NH2·HCl
Quantity 1 g
Formula Weight 201.65
Percent Purity 97%
Grade Technical
Physical Form Crystals or Powder
Chemical Name or Material 2-Amino-4'-methoxyacetophenone hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-4'-methoxyacetophenone hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.