missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-hydroxy-6-methylpyrimidine, 99%

Product Code. 10549790 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10549790 100 g Plastic bottle 100g
10224560 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10549790 Supplier Thermo Scientific Acros Supplier No. 103831000
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3977-29-5 | C5H7N3O | 125.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3977-29-5
Molecular Formula C5H7N3O
Molecular Weight (g/mol) 125.13
MDL Number MFCD00006095
InChI Key KWXIPEYKZKIAKR-UHFFFAOYSA-N
Synonym 2-amino-4-hydroxy-6-methylpyrimidine, 2-amino-6-methylpyrimidin-4-ol, 6-methylisocytosine, mecytosine, 2-amino-6-methylpyrimidin-4 3h-one, superacil, superacyl, 4-methylisocytosine, 2-amino-6-hydroxy-4-methylpyrimidine, 2-amino-6-methyl-4-pyrimidinol
PubChem CID 1532
ChEBI CHEBI:58959
IUPAC Name 2-amino-6-methyl-1H-pyrimidin-4-one
SMILES CC1=CC(=O)N=C(N1)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point >300.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 100 g
Beilstein 24, 343
Solubility Information Solubility in water: insoluble.
Formula Weight 125.13
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Amino-4-hydroxy-6-methylpyrimidine, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.