missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-fluoropyridine, 98%

Product Code. 15421339
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15421339 250 mg 250mg
15401339 1 g 1g
15411339 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15421339 Supplier Thermo Scientific Alfa Aesar Supplier No. H64233.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 944401-77-8 | C5H5FN2 | 112.11 g/mol

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 944401-77-8
Molecular Formula C5H5FN2
Molecular Weight (g/mol) 112.11
MDL Number MFCD09260906
InChI Key WCCFLOQQACDOAX-UHFFFAOYSA-N
Synonym 2-amino-4-fluoropyridine, 4-fluoro-2-pyridinamine, 4-fluoro-2-pyridylamine, 4-fluoro-pyridin-2-ylamine, 4-fluoro-2-pyridineamine, amino-4-fluoropyridine, pubchem19911, acmc-209rrv, 2-pyridinamine,4-fluoro, 4-fluoro-pyridin-2-amine
PubChem CID 18327808
IUPAC Name 4-fluoropyridin-2-amine
SMILES NC1=NC=CC(F)=C1

Specifications

Color Yellow
Quantity 250 mg
Beilstein 15519014
Solubility Information Slightly soluble in water.
Formula Weight 112.04
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Amino-4-fluoropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.