missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chlorobenzonitrile, 99%

Codice prodotto. 15244258 Sfoglia Tutto Thermo Scientific Chemicals Prodotti
Cambia vista
Click to view available options
Quantity:
10 g
50 g
Dimensione della confezione:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
15244258 50 g 50g
15234258 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 15244258 Fornitore Thermo Scientific Acros N. del fornitore 450750500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 38487-86-4 | C7H5ClN2 | 152.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 38487-86-4
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
InChI Key UZHALXIAWJOLLR-UHFFFAOYSA-N
Synonym benzonitrile, 2-amino-4-chloro, 5-chloro-2-cyanoaniline, 4-chloro-2-aminobenzonitrile, 4-chloroanthranilonitrile, 2-amino-4-chlorobenzenecarbonitrile, pubchem20756, 2-amino-4-chloroberizonitrile, bidd:gt0102, 2-amino-4-chlorobenzonitrile, 2-amino-4-chloro-benzonitrile
PubChem CID 170059
IUPAC Name 2-amino-4-chlorobenzonitrile
SMILES C1=CC(=C(C=C1Cl)N)C#N

Specifica

Melting Point 159.0°C to 162.0°C
Quantity 50 g
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Solubility Information Solubility in water: insoluble.
Formula Weight 152.58
Percent Purity 99%
Chemical Name or Material 2-Amino-4-chlorobenzonitrile
compliance-icons
Identificativo prodotto
  • 2-Amino-4-chlorobenzonitrile
Avvertenza
  • Warning
Categoria di pericolo
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Consigli di prudenza
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.