missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloro-6-methylpyrimidine, 98%

Product Code. 11397036
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397036 5 g 5g
11307046 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397036 Supplier Thermo Scientific Alfa Aesar Supplier No. L06896.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5600-21-5 | C5H6ClN3 | 143.574 g/mol

2-Amino-4-chloro-6-methylpyrimidine is a nitrification inhibitor. It was used to study the influence of chlorine substitution in pyrimidine ring on proton donor ability of amino group in 2-aminopyrimidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-chloro-6-methylpyrimidine is a nitrification inhibitor. It was used to study the influence of chlorine substitution in pyrimidine ring on proton donor ability of amino group in 2-aminopyrimidine.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5600-21-5
Molecular Formula C5H6ClN3
Molecular Weight (g/mol) 143.574
MDL Number MFCD00006091
InChI Key NPTGVVKPLWFPPX-UHFFFAOYSA-N
Synonym 2-amino-4-chloro-6-methylpyrimidine, am inhibitor, am pesticide, prepn. am, 4-chloro-6-methyl-2-pyrimidinamine, 4-chloro-6-methylpyrimidin-2-ylamine, 2-pyrimidinamine, 4-chloro-6-methyl, am nitrification inhibitor, pyrimidine, 2-amino-4-chloro-6-methyl, 2-amino-6-chloro-4-methylpyrimidine
PubChem CID 21810
ChEBI CHEBI:58960
IUPAC Name 4-chloro-6-methylpyrimidin-2-amine
SMILES CC1=CC(=NC(=N1)N)Cl

Specifications

Melting Point 181°C to 186°C
Quantity 5 g
Beilstein 114297
Solubility Information Insoluble in water.
Formula Weight 143.58
Percent Purity 98%
Chemical Name or Material 2-Amino-4-chloro-6-methylpyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.