missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloro-5-methylbenzenesulfonic Acid, TRC

Product Code. 17915085
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17915085 1 g 1g
30134022 10 g 10g
30136185 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17915085 Supplier Toronto Research Chemicals Supplier No. TRCA0694951G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-51-7
Formula Weight 220.9913
InChI Formula InChI=1S/C7H8ClNO3S/c1-4-2-7(13(10,11)12)6(9)3-5(4)8/h2-3H,9H2,1H3,(H,10,11,12)
IUPAC Name 2-amino-4-chloro-5-methyl-benzenesulfonic acid
Molecular Formula C7 H8 Cl N O3 S
Molecular Weight (g/mol) 221.66
SMILES Cc1cc(c(N)cc1Cl)S(=O)(=O)O
Synonym 2-Amino-4-chloro-5-methylbenzenesulfonic Acid, 1-Amino-3-chloro-4-methylbenzene-6-sulfonic Acid, 1-Amino-4-methyl-5-chlorobenzene-2-sulfonic Acid, 2-Amino-4-chloro-5-methyl-1-benzenesulfonic Acid, 2-Chloro-4-aminotoluene-5-sulfonic Acid, 2-Chloro-4-toluidine-5-sulfonic Acid, 2-Chloro-p-toluidine-5-sulfonic Acid, 2B Acid, 3-Chloro-4-methylaniline-6-sulfonic Acid, 4-Amino-2-chlorotoluene-5-sulfonic Acid, 4-Amino-6-chlorotoluene-3-sulfonic Acid, 6-Amino-4-chloro-m-toluenesulfonic Acid, Brilliant Toning Red Amine
Chemical Name or Material 2-Amino-4-chloro-5-methylbenzenesulfonic acid

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.