missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-chloro-5,6-dimethylpyrimidine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate.
Solubility
Soluble in Chloroform and Ethanol
Notes
Store at -20°C. Keep away from light and heat.
It is used as a pharmaceutical intermediate.
Solubility
Soluble in Chloroform and Ethanol
Notes
Store at -20°C. Keep away from light and heat.
Specifications
Quantity | 250 mg |
Synonym | 4-chloro-5,6-dimethyl-pyrimidin-2-ylamine, 2-amino-4-chloro-5,6-dimethylpyrimidine, 2-pyrimidinamine, 4-chloro-5,6-dimethyl, 2-amino-4-chloro-5,6-dimethyl-pyrimidine, 6-chloro-4,5-dimethylpyrimidine-2-ylamine, 2-pyrimidinamine,4-chloro-5,6-dimethyl |
Solubility Information | Soluble in Chloroform and Ethanol |
Formula Weight | 157.6 |
Percent Purity | 97% |
Chemical Name or Material | 2-Amino-4-chloro-5,6-dimethylpyrimidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'