missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloro-5,6-dimethylpyrimidine, 97%

Product Code. 15410828
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15410828 5 g 5g
15420828 250 mg 250mg
15400828 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15410828 Supplier Thermo Scientific Alfa Aesar Supplier No. H54552.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14394-61-7 | C6H8ClN3 | 157.601 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in Chloroform and Ethanol

Notes
Store at -20°C. Keep away from light and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14394-61-7
Molecular Formula C6H8ClN3
Molecular Weight (g/mol) 157.601
MDL Number MFCD00462111
InChI Key RRMVDLLCVGZDMH-UHFFFAOYSA-N
PubChem CID 248655
IUPAC Name 4-chloro-5,6-dimethylpyrimidin-2-amine
SMILES CC1=C(N=C(N=C1Cl)N)C

Specifications

Quantity 5 g
Synonym 4-chloro-5,6-dimethyl-pyrimidin-2-ylamine, 2-amino-4-chloro-5,6-dimethylpyrimidine, 2-pyrimidinamine, 4-chloro-5,6-dimethyl, 2-amino-4-chloro-5,6-dimethyl-pyrimidine, 6-chloro-4,5-dimethylpyrimidine-2-ylamine, 2-pyrimidinamine,4-chloro-5,6-dimethyl
Solubility Information Soluble in Chloroform and Ethanol
Formula Weight 157.6
Percent Purity 97%
Chemical Name or Material 2-Amino-4-chloro-5,6-dimethylpyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.