missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloro-3-nitropyridine, 95%

Product Code. 15421989
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15421989 5 g 5g
15431989 250 mg 250mg
15411989 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15421989 Supplier Thermo Scientific Alfa Aesar Supplier No. H64678.06
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 6980-08-1 | C5H4ClN3O2 | 173.56 g/mol

2-Amino-4-chloro-3-nitropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-chloro-3-nitropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in dimethyl formamide, dimethyl sulfoxide, hot ethanol, ethyl acetate and hot methanol.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6980-08-1
Molecular Formula C5H4ClN3O2
Molecular Weight (g/mol) 173.56
MDL Number MFCD04116097
InChI Key DIRINUVNYFAWQF-UHFFFAOYSA-N
Synonym 2-amino-4-chloro-3-nitropyridine, 2-amino-3-nitro-4-chloropyridine, 4-chloro-3-nitro-2-pyridinamine, 4-chloro-3-nitro-pyridin-2-ylamine, 2-pyridinamine, 4-chloro-3-nitro, 4-chloro-3-nitro-pyridin-2-amine, pubchem17739, ksc352m6b, 3-nitro-4-chloropyridine-2-amine, 4-chloro-2-amino-3-nitropyridine
PubChem CID 4071694
IUPAC Name 4-chloro-3-nitropyridin-2-amine
SMILES C1=CN=C(C(=C1Cl)[N+](=O)[O-])N

Specifications

Quantity 5 g
Beilstein 391939
Solubility Information Soluble in dimethyl formamide,dimethyl sulfoxide,hot ethanol,ethyl acetate and hot methanol.
Formula Weight 173.56
Percent Purity 95%
Chemical Name or Material 2-Amino-4-chloro-3-nitropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.