missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-bromobenzoic acid, 97%

Product Code. 15462188
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15462188

Brand: Thermo Scientific Alfa Aesar H55829.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20776-50-5 | C7H6BrNO2 | 216.034 g/mol

2-Amino-4-bromobenzoic acid, , is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-bromobenzoic acid, , is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water (slightly).

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20776-50-5
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD03618454
InChI Key BNNICQAVXPXQAH-UHFFFAOYSA-N
Synonym 4-bromoanthranilic acid, benzoic acid, 2-amino-4-bromo, 5-bromo-2-carboxyaniline, 2-amino-4-bromo-benzoic acid, 2-amino-4-brombenzoic acid, zlchem 336, pubchem13462, acmc-1cb3t, ksc207e6b, pharmabridge p-1344
PubChem CID 88691
IUPAC Name 2-amino-4-bromobenzoic acid
SMILES C1=CC(=C(C=C1Br)N)C(=O)O

Specifications

Melting Point 230°C to 234°C
Quantity 1 g
Solubility Information Soluble in water (slightly).
Formula Weight 216.03
Percent Purity 97%
Chemical Name or Material 2-Amino-4-bromobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.