missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4'-bromoacetophenone hydrochloride, 97%, Thermo Scientific™
Shop All Thermo Scientific Chemicals ProductsSpecifications
Specifications
| CAS Min % | 96.0 |
| CAS Max % | 100.0 |
| Melting Point | 280°C to 285°C |
| Assay Percent Range | 96% min. (HPLC) |
| Packaging | Glass bottle |
| Linear Formula | BrC6H4C(O)CH2NH2·HCl |
| Quantity | 5 g |
| Solubility Information | Solubility in water: soluble |
| Formula Weight | 250.52 |
| Percent Purity | 97% |
| Show More |
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?