missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,5-dimethoxybenzoic Acid, TRC

Product Code. 30323198
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323198 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323198 Supplier Toronto Research Chemicals Supplier No. TRCA60494010G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-4,5-dimethoxybenzoic Acid
CAS 5653-40-7
Molecular Formula C9 H11 N O4
Quantity 10 g
InChI Formula InChI=1S/C9H11NO4/c1-13-7-3-5(9(11)12)6(10)4-8(7)14-2/h3-4H,10H2,1-2H3,(H,11,12)
SMILES COc1cc(N)c(cc1OC)C(=O)O
IUPAC Name 2-amino-4,5-dimethoxybenzoic acid
Molecular Weight (g/mol) 197.19
Formula Weight 197.0688
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.