missing translation for 'onlineSavingsMsg'
Learn More

2'-Amino-4',5'-dimethoxyacetophenone, 98%

Product Code. 11382886
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382886 25 g 25g
11372886 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382886 Supplier Thermo Scientific Alfa Aesar Supplier No. L10605.14
 Restricted Item
Request Bulk or Custom Formats

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4101-30-8 | C10H13NO3 | 195.218 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 4101-30-8
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.218
MDL Number MFCD00016646
InChI Key KGKWXEGYKGTMAK-UHFFFAOYSA-N
Synonym 1-2-amino-4,5-dimethoxyphenyl ethanone, 2'-amino-4',5'-dimethoxyacetophenone, 2-amino-4,5-dimethoxyacetophenone, 1-2-amino-4,5-dimethoxyphenyl ethan-1-one, 1-2-amino-4,5-dimethoxyhenyl ethanone, ethanone, 1-2-amino-4,5-dimethoxyphenyl, 1-acetyl-2-amino-4,5-dimethoxybenzene, pubchem4007, acmc-1arib, 2-acetyl-4,5-dimethoxyaniline
PubChem CID 602085
IUPAC Name 1-(2-amino-4,5-dimethoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1N)OC)OC

Spezifikation

Melting Point 104°C to 108°C
Quantity 25 g
Beilstein 391380
Formula Weight 195.22
Percent Purity 98%
Chemical Name or Material 2'-Amino-4',5'-dimethoxyacetophenone

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.