missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,5-difluorobenzoic acid, 97%

Product Code. 11354316
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354316 25 g 25g
11339965 1 g 1g
11349965 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11354316 Supplier Thermo Scientific Alfa Aesar Supplier No. L19950.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 83506-93-8 | C7H5F2NO2 | 173.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83506-93-8
Molecular Formula C7H5F2NO2
Molecular Weight (g/mol) 173.12
MDL Number MFCD00077509
InChI Key DGOZIZVTANAGCA-UHFFFAOYSA-N
Synonym 4,5-difluoroanthranilic acid, 2-amino-4,5-difluoro-benzoic acid, buttpark 49\07-41, benzoicacid, 2-amino-4,5-difluoro, benzoic acid, 2-amino-4,5-difluoro, pubchem4675, 2-amino-4,5-difluoro, 4,5-difluoroantranilic acid, ksc448a3l, 4,5-difluoro anthranilic acid
PubChem CID 735965
IUPAC Name 2-amino-4,5-difluorobenzoic acid
SMILES NC1=CC(F)=C(F)C=C1C(O)=O

Specifications

Melting Point 181°C to 183°C
Quantity 25 g
Beilstein 2834162
Formula Weight 173.12
Percent Purity 97%
Chemical Name or Material 2-Amino-4,5-difluorobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.