missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4,5-dichloropyrimidine, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2-Amino-4,6-dichloropyrimidine |
| CAS | 56-05-3 |
| Molecular Formula | C4 H3 Cl2 N3 |
| Quantity | 5 g |
| Synonym | 2-Amino-4,6-Dichloropyrimidine |
| InChI Formula | InChI=1S/C4H3Cl2N3/c5-2-1-3(6)9-4(7)8-2/h1H,(H2,7,8,9) |
| SMILES | Nc1nc(Cl)cc(Cl)n1 |
| IUPAC Name | 4,6-dichloropyrimidin-2-amine |
| Molecular Weight (g/mol) | 163.99 |
| Formula Weight | 162.9704 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction