missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile, 97%

Product Code. 15448125
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448125 10 g 10g
15438125 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448125 Supplier Thermo Scientific Alfa Aesar Supplier No. H32544.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4651-91-6 | C9H10N2S | 178.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 4651-91-6
Molecular Formula C9H10N2S
Molecular Weight (g/mol) 178.253
MDL Number MFCD00128278
InChI Key ADHVMGAFAKSNOM-UHFFFAOYSA-N
Synonym 2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carbonitrile, 2-amino-3-cyano-4,5,6,7-tetrahydrobenzo b thiophene, 2-amino-4,5,6,7-tetrahydro-benzo b thiophene-3-carbonitrile, benzo b thiophene-3-carbonitrile, 2-amino-4,5,6,7-tetrahydro, 2-amino-4,5,6,7-tetrahydro-1-benzo b thiophene-3-carbonitrile, 2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carbonitryle, cambridge id 5130181, aminotetrahydrobenzothiophenecarbonitrile
PubChem CID 78382
IUPAC Name 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile
SMILES C1CCC2=C(C1)C(=C(S2)N)C#N

Spezifikation

Melting Point 141°C to 144°C
Quantity 10 g
Formula Weight 178.26
Percent Purity 97%
Physical Form Crystals or Powder or Crystalline Powder
Chemical Name or Material 2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.