missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(2-naphthyl)thiazole, 98%, Thermo Scientific Chemicals

Product Code. 11490827
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21331-43-1
Molecular Formula C13H10N2S
Molecular Weight (g/mol) 226.297
MDL Number MFCD00046452
InChI Key GWDNDNTTXIIXRS-UHFFFAOYSA-N
Synonym 4-naphthalen-2-yl-thiazol-2-ylamine, 4-naphthalen-2-yl-1,3-thiazol-2-amine, 4-2-naphthyl-1,3-thiazol-2-amine, 2-amino-4-2-naphthyl thiazole, 4-naphthalen-2-yl thiazol-2-amine, 4-naphthalen-2-yl-thiazol-2-yl-amine, 4-2-naphthalenyl-2-thiazolamine, 2-thiazolamine,4-2-naphthalenyl, 4-2-naphthyl-1,3-thiazole-2-ylamine, acmc-20ao57
PubChem CID 673702
IUPAC Name 4-naphthalen-2-yl-1,3-thiazol-2-amine
SMILES C1=CC=C2C=C(C=CC2=C1)C3=CSC(=N3)N

Specifications

Melting Point 153°C to 156°C
Quantity 1 g
Beilstein 173533
Formula Weight 226.3
Percent Purity 98%
Chemical Name or Material 2-Amino-4-(2-naphthyl)thiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'