missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-(trifluoromethyl)pyridine-5-boronic acid pinacol ester, 96%

Product Code. 15421606
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15421606 250 mg 250mg
15411606 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15421606 Supplier Thermo Scientific Alfa Aesar Supplier No. H54793.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 947249-01-6 | C12H16BF3N2O2 | 288.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 947249-01-6
Molecular Formula C12H16BF3N2O2
Molecular Weight (g/mol) 288.08
MDL Number MFCD12923414
InChI Key OQZKROYNCVLJKM-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3-trifluoromethyl pyridin-2-amine, 2-amino-3-trifluoromethyl pyridine-5-boronic acid pinacol ester, 6-amino-5-trifluoromethyl pyridine-3-boronic acid pinacol ester, 6-amino-5-trifluoromethyl pyridin-3-yl boronic acid pinacol ester, 3-trifluoromethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, 5-4,4,5,5-tetra methyl-1,3,2 dioxaborolan-2-yl-3-trifluoromethyl-pyridin-2-ylamine, 5-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl-3-trifluoromethyl-2-pyridylamine, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3-trifluoromethyl-pyridin-2-ylamine, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3-trifluoromethyl pyridin-2-amine, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3-trifluoromethyl-pyridin-2-yl-amine
PubChem CID 46864103
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)pyridin-2-amine
SMILES CC1(C)OB(OC1(C)C)C1=CC(=C(N)N=C1)C(F)(F)F

Specifications

Melting Point 132°C to 134°C
Quantity 250 mg
Formula Weight 288.07
Percent Purity 96%
Chemical Name or Material 2-Amino-3-(trifluoromethyl)pyridine-5-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.