missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-amino-3-pyridinecarboxaldehyde 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 95.0°C to 102.0°C |
Color | Yellow to Brown |
Flash Point | >300°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Quantity | 1 g |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in chloroform |
Show More |
Safety and Handling
Product Identifier
- 2-Amino-3-pyridinecarboxaldehyde
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'