missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-picoline, 96%

Product Code. 10376400 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10376400 100 g Glass bottle 100g
10469390 5 g Glass bottle 5g
10714211 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10376400 Supplier Thermo Scientific Acros Supplier No. 104381000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1603-40-3 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1603-40-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006320
InChI Key RGDQRXPEZUNWHX-UHFFFAOYSA-N
Synonym 2-amino-3-picoline, 2-amino-3-methylpyridine, 2-pyridinamine, 3-methyl, 3-methyl-2-pyridinamine, 3-methyl-2-aminopyridine, 3-methyl-2-pyridylamine, 3-picoline, 2-amino, 2-amino-3-methyl pyridine, 2-amino-beta-picoline, pyridine, 2-amino-3-methyl
PubChem CID 15347
IUPAC Name 3-methylpyridin-2-amine
SMILES CC1=C(N=CC=C1)N

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 32.0°C to 34.0°C
Color Yellow
Boiling Point 221.0°C to 222.0°C
Flash Point 111°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5813 to 1.5833
Quantity 100 g
Beilstein 22, V,9, 272
Solubility Information Solubility in water: freely soluble.
Formula Weight 108.14
Percent Purity 96%
Physical Form Liquid After Melting
Chemical Name or Material 2-Amino-3-picoline, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-3-picoline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.