missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3'-nitroacetophenone hydrochloride, 98%

Product Code. 11444873
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444873 1 g 1g
11454873 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444873 Supplier Thermo Scientific Alfa Aesar Supplier No. A12205.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 36765-84-1 | C8H9N2O3 | 181.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36765-84-1
Molecular Formula C8H9N2O3
Molecular Weight (g/mol) 181.17
MDL Number MFCD00051997
InChI Key NXCCDPOCDBAGFX-UHFFFAOYSA-O
Synonym 3-nitrophenacylamine hydrochloride, 2-amino-1-3-nitrophenyl ethanone hydrochloride, 2-amino-3'-nitroacetophenone hydrochloride, 2-amino-1-3-nitrophenyl ethan-1-one hydrochloride, acmc-20amzi, 2-amine-3'-nitroacetophenone hcl, 2-amino-1-3-nitrophenyl ethanone hcl salt, 2-amino-3-nitroacetophenone hydrochloride
PubChem CID 2774374
IUPAC Name 2-amino-1-(3-nitrophenyl)ethanone;hydrochloride
SMILES [NH3+]CC(=O)C1=CC=CC(=C1)[N+]([O-])=O

Specifications

Melting Point 210°C to 212°C (decomposition)
Quantity 1 g
UN Number UN1759
Formula Weight 216.62
Percent Purity 98%
Chemical Name or Material 2-Amino-3'-nitroacetophenone hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.