missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-nitro-4-picoline, 98%, Thermo Scientific™

Product Code. 10701545 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10701545 25 g Glass bottle 25g
10123641 1 g Glass bottle 1g
10043610 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10701545 Supplier Thermo Scientific Acros Supplier No. 319550250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6635-86-5
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.14
MDL Number MFCD00006315
InChI Key IKMZGACFMXZAAT-UHFFFAOYSA-N
Synonym 2-amino-4-methyl-3-nitropyridine, 2-amino-3-nitro-4-picoline, 2-pyridinamine, 4-methyl-3-nitro, 2-amino-3-nitro-4-methylpyridine, 4-methyl-3-nitro-pyridin-2-ylamine, 4-methyl-3-nitro-2-pyridylamine, 4-methyl-3-nitro-2-pyridinamine, 4-methyl-3-nitro-2-aminopyridine, 4-methyl-3-nitropyridin-2-amin, 4mrz
PubChem CID 243166
IUPAC Name 4-methyl-3-nitropyridin-2-amine
SMILES CC1=C(C(=NC=C1)N)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 136°C to 141°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 153.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-nitro-4-picoline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.