missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-methylpyridine, 97%

Product Code. 11319699
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319699 25 g 25g
11463913 100 g 100g
11473913 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11319699 Supplier Thermo Scientific Alfa Aesar Supplier No. A10224.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1603-40-3 | C6H8N2 | 108.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1603-40-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006320
InChI Key RGDQRXPEZUNWHX-UHFFFAOYSA-N
Synonym 2-amino-3-picoline, 2-amino-3-methylpyridine, 2-pyridinamine, 3-methyl, 3-methyl-2-pyridinamine, 3-methyl-2-aminopyridine, 3-methyl-2-pyridylamine, 3-picoline, 2-amino, 2-amino-3-methyl pyridine, 2-amino-beta-picoline, pyridine, 2-amino-3-methyl
PubChem CID 15347
IUPAC Name 3-methylpyridin-2-amine
SMILES CC1=C(N=CC=C1)N

Specifications

Melting Point 29°C to 32°C
Density 1.073
Boiling Point 221°C to 222°C
Flash Point 118°C (244°F)
Odor Unpleasant
Refractive Index 1.5825
Quantity 25 g
UN Number UN2811
Beilstein 107892
Sensitivity Hygroscopic
Formula Weight 108.14
Percent Purity 97%
Chemical Name or Material 2-Amino-3-methylpyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.