missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-3-methylpentan-1-ol, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 2-Amino-3-methylpentan-1-ol |
| CAS | 24629-25-2 |
| Molecular Formula | C6H15NO |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C6H15NO/c1-3-5(2)6(7)4-8/h5-6,8H,3-4,7H2,1-2H3/t5-,6+/m0/s1 |
| SMILES | CC[C@H](C)[C@H](N)CO |
| IUPAC Name | (2S,3S)-2-amino-3-methylpentan-1-ol |
| Molecular Weight (g/mol) | 117.19 |
| Formula Weight | 117.1154 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction