missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-methylbenzoic acid, 98%

Product Code. 11461267
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461267 5 g 5g
11451267 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11461267 Supplier Thermo Scientific Alfa Aesar Supplier No. B24230.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4389-45-1 | C8H9NO2 | 151.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4389-45-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00007745
InChI Key WNAJXPYVTFYEST-UHFFFAOYSA-N
Synonym 3-methylanthranilic acid, 2-amino-3-methyl-benzoic acid, 2-amino-3-methylbenzoicacid, 3-methyl-2-aminobenzoic acid, benzoic acid, 2-amino-3-methyl, 2-amino-m-toluic acid, 2-amino-3-methyl benzoic acid, m-toluic acid, 2-amino, unii-8rgx6sqe9n, 2-amino-3-methylbenzoate
PubChem CID 78101
ChEBI CHEBI:80574
IUPAC Name 2-amino-3-methylbenzoic acid
SMILES CC1=CC=CC(=C1N)C(=O)O

Specifications

Melting Point 174°C to 177°C
Quantity 5 g
Beilstein 2359694
Formula Weight 151.16
Percent Purity 98%
Chemical Name or Material 2-Amino-3-methylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.