missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-methylbenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 10264270 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10264270 5 g Glass bottle 5g
10549610 10 g Glass bottle 10g
10015903 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10264270 Supplier Thermo Scientific Acros Supplier No. 103970050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4389-45-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.16
MDL Number MFCD00007745
InChI Key WNAJXPYVTFYEST-UHFFFAOYSA-N
Synonym 3-methylanthranilic acid, 2-amino-3-methyl-benzoic acid, 2-amino-3-methylbenzoicacid, 3-methyl-2-aminobenzoic acid, benzoic acid, 2-amino-3-methyl, 2-amino-m-toluic acid, 2-amino-3-methyl benzoic acid, m-toluic acid, 2-amino, unii-8rgx6sqe9n, 2-amino-3-methylbenzoate
PubChem CID 78101
ChEBI CHEBI:80574
IUPAC Name 2-amino-3-methylbenzoic acid
SMILES CC1=CC=CC(=C1N)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 173.0°C to 177.0°C
Color Brown-Gray to Pink
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula H2NC6H3(CH3)CO2H
Quantity 5 g
Beilstein 14, 479
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 151.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-methylbenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-3-methylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.