missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-methoxybenzoic acid

Product Code. p-7042899
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
This item is not returnable. View return policy

Product Code. 15456279

Brand: Thermo Scientific Alfa Aesar J64244.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3177-80-8 | C8H9NO3 | 167.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3177-80-8
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00075178
InChI Key SXOPCLUOUFQBJV-UHFFFAOYSA-N
Synonym 3-methoxyanthranilic acid, benzoic acid, 2-amino-3-methoxy, 3-methoxyanthranilate, m-anisic acid, 2-amino, 3-methoxy-2-aminobenzoic acid, 2-amino-3-methoxy-benzoic acid, 2-amino-3-methoxy benzoic acid, 2-amino-3-methyloxy benzoic acid, 2-amino-3-methoxybenzoicacid
PubChem CID 255720
ChEBI CHEBI:27440
IUPAC Name 2-amino-3-methoxybenzoic acid
SMILES COC1=CC=CC(=C1N)C(=O)O

Specifications

Melting Point 169°C to 170°C
Color White
Quantity 5 g
Formula Weight 167.16
Physical Form Powder
Chemical Name or Material 2-Amino-3-methoxybenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.