missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-hydroxypyridine, 98%

Product Code. 11329699
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329699 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11329699 Supplier Thermo Scientific Alfa Aesar Supplier No. A10665.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16867-03-1 | C5H6N2O | 110.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 16867-03-1
Melting Point 166°C to 174°C
Odor Odorless
Molecular Formula C5H6N2O
MDL Number MFCD00006317
Quantity 100 g
UN Number UN2811
Beilstein 109868
Synonym 2-amino-3-hydroxypyridine, 2-amino-3-pyridinol, 3-pyridinol, 2-amino, 2-amino-3-pyridol, 3-hydroxy-2-pyridinamine
InChI Key BMTSZVZQNMNPCT-UHFFFAOYSA-N
SMILES NC1=NC=CC=C1O
IUPAC Name 2-aminopyridin-3-ol
Molecular Weight (g/mol) 110.12
PubChem CID 28114
Formula Weight 110.12
Percent Purity 98%
Chemical Name or Material 2-Amino-3-hydroxypyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.