missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog Number 15486946
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1831130-33-6
2-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-amino-3-fluorophenylboronic acid pinacol ester, 2-amino-3-fluorobenzeneboronic acid pinacol ester, 2-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine
This item is not returnable. View return policy
1831130-33-6
2-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-amino-3-fluorophenylboronic acid pinacol ester, 2-amino-3-fluorobenzeneboronic acid pinacol ester, 2-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 237.08
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Amino-3-fluorobenzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'