missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-cyano-4,5-di(fur-2-yl)furan, 97%, Thermo Scientific™

Product Code. 10621683
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10621683 250 mg 250mg
10314722 1 g 1g
10332255 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10621683 Supplier Thermo Scientific Maybridge Supplier No. NRB00495CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 24386-17-2
Molecular Formula C13H8N2O3
Molecular Weight (g/mol) 240.22
MDL Number MFCD00099027
InChI Key RDQSOEAGWZLHBE-UHFFFAOYSA-N
Synonym 2-amino-3-cyano-4,5-di fur-2-yl furan, 5'-amino-2,2':3',2-terfuran-4'-carbonitrile, 2-amino-3-cyano-4,5-di furyl furan, 2,2':3',2-terfuran-4'-carbonitrile,5'-amino, 2-amino-4,5-bis furan-2-yl furan-3-carbonitrile, 5'-amino-2'-furan-2-yl-2,3'-bifuran-4'-carbonitrile, 2-amino-4,5-di 2-furyl furan-3-carbonitrile, 5-amino-3-furan-2-yl-2,2'-bifuran-4-carbonitrile, aminocyanodifurylfuran
PubChem CID 714249
SMILES NC1=C(C#N)C(C2=CC=CO2)=C(O1)C1=CC=CO1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 5'-amino-2'-(furan-2-yl)-[2,3'-bifuran]-4'-carbonitrile
Formula Weight 240.22
Percent Purity 97%
Chemical Name or Material 2-Amino-3-cyano-4, 5-di(fur-2-yl)furan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.