missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-chlorobenzonitrile, 97%

Product Code. 11377529
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377529 5 g 5g
11367529 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377529 Supplier Thermo Scientific Alfa Aesar Supplier No. H27918.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 53312-77-9 | C7H5ClN2 | 152.581 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53312-77-9
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.581
MDL Number MFCD03425881
InChI Key FAHVRFGAGJMXHW-UHFFFAOYSA-N
Synonym benzonitrile, 2-amino-3-chloro, pubchem4612, acmc-20a0rc, amino-3-chlorobenzonitrile, ksc494a6d, 2-amino-3-chloro-benzonitrile, 3-chloroanthranilonitrile, 2-amino-3-chlorobenzenecarbonitrile
PubChem CID 12627363
IUPAC Name 2-amino-3-chlorobenzonitrile
SMILES C1=CC(=C(C(=C1)Cl)N)C#N

Specifications

Melting Point 94°C to 96°C
Quantity 5 g
UN Number UN3439
Formula Weight 152.58
Percent Purity 97%
Chemical Name or Material 2-Amino-3-chlorobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.